missing translation for 'onlineSavingsMsg'
Learn More
Mutant IDH1-IN-1, MedChemExpress Product Code.: 16425198

Mutant IDH1-IN-1, MedChemExpress

Product Code. 16425198
10 mg, 10mg
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16425198

Brand: Medchem Express HY12475

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Mutant IDH1-IN-1 is a mutant-selective IDH1 inhibitor with with IC50s of 4, 42, 80 and 143 nM against mutant IDH1 R132C/R132C, IDH1 R132H/R132H, IDH1 R132H/WT and wild type IDH1, respectively.

Mutant IDH1-IN-1 is a mutant-selective IDH1 inhibitor with with IC50s of 4, 42, 80 and 143 nM against mutant IDH1 R132C/R132C, IDH1 R132H/R132H, IDH1 R132H/WT and wild type IDH1, respectively.

Specifications

Chemical Name or Material Mutant IDH1-IN-1
CAS 1355326-21-4
Color White
Purity Grade Notes Research
Molecular Formula C30H31FN4O2
Quantity 10 mg
Solubility Information DMSO : ≥ 45 mg/mL (90.25 mM)
SMILES O=C(C(C1=C(C)C=CC=C1)N(C(CN2C=NC3=C2C=CC=C3)=O)C4=CC(F)=CC=C4)NC5CCCCC5
Molecular Weight (g/mol) 498.59
Formula Weight 498.59
Percent Purity 99.53%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Mutant IDH1-IN-1, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.