Learn More
ZVG™ Multitex™ Premium Extra z80 Cleaning Cloth Roll
Product Code. 17823525
Click to view available options
Color:
Blue
White
Unit Size:
1 roll
Product Code. 17823525
Brand: ZVG™ 1114280B
Only null left
Add to basket
Add to basket
This cloth roll provides extra thick cleaning wipes made of very soft, highly absorbent viscose nonwoven material.
  • High quality, nonwoven fabrics suitable for both wet and dry cleaning due to their tremendous strength
  • Very soft and lint-free
  • Able to absorb large amounts of liquid while remaining tear-resistant and solvent-resistant
  • Multitex™ cloths can be washed and wrung out during cleaning
  • Frequently used in sensitive work areas such as precision mechanics, painting shops, the electrical and automotive industries, as well as in nursing homes
  • Similar to textile cleaning rags, and can be used for different durations depending on level of contamination
  • Viscose non-woven fabric with textile characteristics
  • Solvent-resistant
  • Silicone- and lint-free
  • Roll contains 475 tear-off sheets
Viscose fibers are industrially produced from the base material cellulose. The chemical composition of this 'natural synthetic fiber' is similar to that of cotton. Viscose products are absorbent, flexible, and durable due to their textile character.
Specifications
Product Type Cleaning Cloth
Length (English) 15 in.
Length (Metric) 38 cm
Width (English) 12.6 in.
Width (Metric) 32 cm
Material Polyester, Viscose
Color Blue