Learn More
ZVG™ Multisoft™ Air+ Wipes
These lint-free, highly absorbent, wipes are made of soft embossed white cellulose.
Brand:  ZVG™ 10104-00
 View more versions of this product
Product Code. 17883665
  • 1327.97 NOK / Pack of 500
Estimated Shipment: 24-10-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Sensitive surfaces require gentle treatment. Multisoft™ extra soft, low-lint specialty cloths are an ideal choice for care and polishing tasks. They efficiently absorb liquids, such as polishing wax, and evenly release it without compromising their strength. In the zetPutz™ range, you will find additional low-lint, highly absorbent 1- to 4-ply cleaning and polishing cloths for various cleaning purposes.
  • Airlaid material
  • Highly absorbent
  • Embossed
  • Latex- and silicone-free
  • Solvent-resistant
Specifications
Specifications
Wipe
32 cm
34 cm
White
500 Cloths
12.6 in.
13.4 in.
Fleece
Single-use
Non-sterile