Learn More
IKA™ Multi-use Milling Tube MMT 40.1
Durable, dishwasher-safe grinding chamber/milling tube.
Marque:  IKA™ 0020003165
2987.75 NOK valid until 2024-07-30
Use promo code "21657" to get your promotional price.
Informations supplémentaires : Weight : 1.70000kg
Code produit. 15413501
  • 3145.00 NOK / 1 set
Estimated Shipment: 23-07-2024
to see stock.
Ajouter au panier
Description
Description
  • Can be used and cleaned in a dishwasher multiple times
  • Package includes a rich set of spare parts so that wearing parts can be replaced if necessary
  • Delivery: 1 x MMT 40, 25 x sealing, 5 x beater, 5 x coupling
  • Additional abrasion set A-MMT 40.100 available
Spécification
Spécification
Milling Tube, Grinding Chamber
40 mL
Rich set of spare parts, 25 x sealings, 5 x beaters, 5 x couplings
Suggestions de produits
Suggestions de produits
Vidéos
FDS
Documents
Documents
Certificats