Learn More
Novus Biologicals™ Multi-Species Brain Tissue MicroArray (Normal)
The panel contains 5 to 8 different tissues on one slide.
Brand: Novus Biologicals™ NBP2-78096
This item is not returnable.
View return policy
Description
Specifications
5 Slides | |
Multi-species | |
Please bake slides at 60C for 30 minutes before use. |
Product Content Correction
Product Title
Novus Biologicals™ Multi-Species Brain Tissue MicroArray (Normal) > 5 Slides