Learn More
Novus Biologicals™ Multi-Species Brain Tissue MicroArray (Normal)
The panel contains 5 to 8 different tissues on one slide.
Brand:  Novus Biologicals™ NBP2-78096
Product Code. 18673041
  • 6245.00 NOK / Pack of 5
Specifications
5 Slides
Multi-species
Please bake slides at 60C for 30 minutes before use.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Novus Biologicals™ Multi-Species Brain Tissue MicroArray (Normal) > 5 Slides