Learn More
Nabertherm™ Muffle Furnace With Lift Door, 1100°C
Muffle furnaces are the reliable and long-lasting all-rounders in the laboratory and are ideally suited for a large number of processes in the field of material research and heat treatment.
46290.00 NOK - 100130.00 NOK
Specifications
Temperature (Metric) | 1100°C |
---|---|
Type | Furnace |
Style | Lift Door |
Frequency | 50/60 Hz |
Product Code | Brand | Capacity (Metric) | Controller Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Controller Type | Price | Quantity & Availability | |||||
17347849
|
Nabertherm™
L-035H2ANL |
3 L | B510 |
46290.00 NOK
Each |
Estimated Shipment: 04-07-2025
Log in to see stock. |
|||||
17357849
|
Nabertherm™
L-035H2ANO |
3 L | C550 |
55525.00 NOK
Each |
Estimated Shipment: 04-07-2025
Log in to see stock. |
|||||
17327859
|
Nabertherm™
L-055H2ANL |
5 L | B510 |
55760.00 NOK
Each |
Estimated Shipment: 04-07-2025
Log in to see stock. |
|||||
17337859
|
Nabertherm™
L-055H2ANO |
5 L | C550 |
65000.00 NOK
Each |
Estimated Shipment: 04-07-2025
Log in to see stock. |
|||||
17387869
|
Nabertherm™
L-095H2ANL |
9 L | B510 |
64280.00 NOK
Each |
Estimated Shipment: 04-07-2025
Log in to see stock. |
|||||
17397869
|
Nabertherm™
L-095H2ANO |
9 L | C550 |
73510.00 NOK
Each |
Estimated Shipment: 04-07-2025
Log in to see stock. |
|||||
17347889
|
Nabertherm™
L-155H2ANL |
15 L | B510 |
67280.00 NOK
Each |
Estimated Shipment: 04-07-2025
Log in to see stock. |
|||||
17357889
|
Nabertherm™
L-155H2ANO |
15 L | C550 |
76510.00 NOK
Each |
Estimated Shipment: 04-07-2025
Log in to see stock. |
|||||
17307909
|
Nabertherm™
L-245H2ANL |
24 L | B510 |
90900.00 NOK
Each |
Estimated Shipment: 04-07-2025
Log in to see stock. |
|||||
17317909
|
Nabertherm™
L-245H2ANO |
24 L | C550 |
100130.00 NOK
Each |
Estimated Shipment: 04-07-2025
Log in to see stock. |
|||||
17387909
|
Nabertherm™
L-405H2ANL |
40 L | B510 |
95215.00 NOK
Each |
Estimated Shipment: 04-07-2025
Log in to see stock. |
|||||
17397909
|
Nabertherm™
L-405K1ANL |
40 L | C550 |
82745.00 NOK
Each |
Estimated Shipment: 04-07-2025
Log in to see stock. |
|||||
Description
- Optimal temperature uniformity of +/- 5K with closed fresh air inlet in empty workspace
- Dual shell housing made of textured stainless steel sheets with additional fan cooling for low surface temperature
- Only fiber materials are used which are not classified as carcinogenic according to TRGS 905, class 1 or 2
- Ceramic heating plates with integral heating elements which are safeguarded and easy to replace
- Adjustable air inlet integrated in door
- Exhaust air outlet in rear wall of furnace
- Solid state relays provide for low-noise operation
- NTLog Basic for Nabertherm controller: recording of process data with USB-flash drive
- Defined application within the constraints of the operating instructions
- Very short delivery times for standard furnaces and spare parts
Additional equipment
- Chimney, chimney with fan or catalytic converter
- Over-temperature limit controller with adjustable cutout temperature for thermal protection
- Protective gas connection to purge with non-flammable gases
- Manual or automatic gas supply system
- Port for thermocouple in the rear wall or in the furnace door
- Process control and documentation via VCD software package for monitoring, documentation and control
Specifications
1100°C | |
Furnace | |
Lift Door | |
50/60 Hz |
Product Title
Nabertherm™ Muffle Furnace With Lift Door, 1100°C