Learn More
MTHFR Monoclonal Antibody (5D3), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA515844
Description
MA5-15844 targets MTHFR in indirect ELISA, IHC and WB applications and shows reactivity with Human and Rat samples. The MA5-15844 immunogen is purified recombinant fragment of human MTHFR expressed in E. Coli. . MA5-15844 detects MTHFR which has a predicted molecular weight of approximately 75kDa.
The protein encoded by this gene catalyzes the conversion of 5,10-methylenetetrahydrofolate to 5-methyltetrahydrofolate, a co-substrate for homocysteine remethylation to methionine. Genetic variation in this gene influences susceptibility to occlusive vascular disease, neural tube defects, colon cancer and acute leukemia, and mutations in this gene are associated with methylenetetrahydrofolate reductase deficiency.
Specifications
MTHFR | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
P42898 | |
MTHFR | |
Purified recombinant fragment of human MTHFR expressed in E. Coli. | |
RUO | |
362657, 4524 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
5D3 | |
Unconjugated | |
MTHFR | |
5,10-methylenetetrahydrofolate reductase; 5,10-methylenetetrahydrofolate reductase (NADPH); AI323986; Methylenetetrahydrofolate reductase; methylenetetrahydrofolate reductase (NAD(P)H); MTHFR; RP11-56N19.4 | |
Mouse | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.