missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTCO2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55070-1-AP-150UL
This item is not returnable.
View return policy
Description
The antibody is specific to COX2.
Cytochrome c oxidase is the fourth complex in the respiratory chain and is responsible for catalyzing the conversion of O2 to H2O. Subunit 2 of the cytochrome c oxidase complex combines with two other subunits (1 and 3) to form a core protein structure that performs many functions of the enzyme. The metallic copper center of this subunit transfers electrons to the heme center of subunit 1, which results in the movement of electrons from cytochrome c to the heme A3 and copper B metallic center of complex IV. Defects in subunit 2 of cytochrome c oxidase can result in COX deficiency, which causes a wide range of symptoms from local myopathy to multiple system pathologies that begin between infancy and adulthood. Abnormalities in this subunit are also associated with tumor development.Specifications
MTCO2 | |
Polyclonal | |
Unconjugated | |
COX2 | |
COII, COX2, COXII, cytochrome c oxidase II, Cytochrome c oxidase subunit 2, MT CO2, MTCO2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
17709, 26198, 4513 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P00403, P00405 | |
COX2 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction