missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSH6 Mouse anti-Human, Clone: 2E10B2, Proteintech
Mouse Monoclonal Antibody
1696.32 NOK - 4796.50 NOK
Specifications
Antigen | MSH6 |
---|---|
Clone | 2E10B2 |
Concentration | 1.03 mg/mL |
Applications | Immunofluorescence, Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16847805
|
Proteintech
66172-1-IG-20UL |
20 μL |
1696.32 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16837805
|
Proteintech
66172-1-IG-150UL |
150 μL |
4796.50 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The DNA mismatch repair MSH2/MSH6 heterodimer is responsible for the detection of DNA damage, and may promote apoptosis in normal cells, thus protecting mammals from ultraviolet-induced maligt transformation and hence the prevention of skin cancer.Specifications
MSH6 | |
1.03 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
G/T mismatch binding protein, GTBP, GTMBP, hMSH6, HNPCC5, HSAP, MSH6, MutS alpha 160 kDa subunit, mutS homolog 6 (E. coli), p160 | |
MSH6 | |
IgG2a | |
Protein A | |
Antibody |
2E10B2 | |
Immunofluorescence, Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry | |
Unconjugated | |
Mouse | |
Human | |
P52701 | |
2956 | |
MSH6 Fusion Protein Ag12645 | |
Primary | |
-20°C | |
MSH6 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title