Learn More
STEMCELL Technologies MSC Characterization Antibody Panel
Multi-color flow cytometry kit for characterization of human mesenchymal stromal cells
Brand:  STEMCELL Technologies 100-0354
Encompass_Preferred
Product Code. 17177571
  • 9334.09 NOK / 1 set
Description
  • The MSC Characterization Antibody Panel is designed for the phenotypic analysis of human mesenchymal stromal cells (MSCs)
  • This kit contains three fluorochrome-conjugated antibodies for the identification of human MSCs, and a fluorochrome-conjugated antibody for the identification of non-MSCs
  • When measured by flow cytometry, ≥ 95% of the MSC population must express CD73, CD90, and CD105, and these cells must lack expression (≤ 2% positive) of CD45
  • These reagents provide single-step labeling for the verification of human MSCs
  • The MSC Characterization Antibody Panel can be used to identify human MSCs derived from various tissues such as bone marrow MSCs, pluripotent stem cell-derived mesenchymal progenitor cells, adipose-derived MSCs, dental pulp-derived MSCs, and umbilical cord-derived MSCs
Specifications
Specifications
Flow Cytometry
Monoclonal
1 Kit
Mesenchymal Stromal Cells
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
STEMCELL Technologies MSC Characterization Antibody Panel