missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ T-Butylated Hyrdoquinone
Purity: 99%
Brand: MP Biomedicals™ 0296037480
This item is not returnable.
View return policy
Description
Molecular Formula: C10H14O2
Molecular Weight: 166.22
CAS No.: 1948-33-0
Synonym: BHQ; 2,5-Di-tert-butylhydroxyquinone; 2-(1,1-Dimethylethyl)-1,4-Benzenediol
Physical Description: White to pink crystalline powder
Description: CAO-3 Antioxidant is the food grade desgnation for 2,6-di-tert-butyl-para-cresol (BHT Butylated Hydroxytoluene). CAO-3 is a direct replacement for CAO-1 (technical grade) BHT
Solubility: Soluble in alcohol, ether and methanol, acetone; slightly soluble in water (0.5%)
Molecular Weight: 166.22
CAS No.: 1948-33-0
Synonym: BHQ; 2,5-Di-tert-butylhydroxyquinone; 2-(1,1-Dimethylethyl)-1,4-Benzenediol
Physical Description: White to pink crystalline powder
Description: CAO-3 Antioxidant is the food grade desgnation for 2,6-di-tert-butyl-para-cresol (BHT Butylated Hydroxytoluene). CAO-3 is a direct replacement for CAO-1 (technical grade) BHT
Solubility: Soluble in alcohol, ether and methanol, acetone; slightly soluble in water (0.5%)
Specifications
T-Butylated Hyrdoquinone | |
100 g | |
2°C to 8°C |
Powder | |
Swine |