missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Streptokinase
Catalyzes hydrolysis of amide linkages
Brand: MP Biomedicals™ 0210111450_CLEAN
This item is not returnable.
View return policy
Description
Forms a 1:1 complex with plasminogen which is then converted to streptokinase-plasmin. Both streptokinase-plasminogen and streptokinase-plasmin complexes can activate plasminogen.
- Can be used as a thrombolytic agent in therapy of ischemic stroke
- From: Lancefield group C strain of Streptococcus
- Presentation: White lyophilized powder
- Unit definition: Amount of enzyme that will liquefy a standard clot of fibrinogen, plasminogen and thrombin in 10 minutes at 37°C and pH 7.5
- Specific activity: >5,000 units/mg dry weight
- Solubility: Soluble in water
Specifications
≥5000 U/mg | |
Used as a thrombolytic agent in the therapy of ischemic stroke | |
Streptococcus species, lancefield group C strain | |
Streptokinase | |
Lyophilized Powder |
White | |
-20°C | |
50 kU | |
47400 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction