Learn More
MP Biomedicals Îł-Globulin, Mouse Purified Product Code.: 11474235
MP Biomedicals Îł-Globulin, Mouse Purified
Product Code. 11474235 Shop All MP Biomedicals Products
25 mg, 25mg
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Product Code. 11474235
Brand: MP Biomedicals 0855861
Specifications
Concentration 20 mg/mL
Quantity 25 mg
Content And Storage 2°C to 8°C
Form Lyophilized
For Use With (Application) Immunoassay
Product Type Purified Mouse gamma Globulin
Product Content Correction
Product Title
MP Biomedicals Îł-Globulin, Mouse Purified >