Learn More
MP Biomedicals™ γ-Globulin, Human Purified
Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
Concentration | 17mg/mL |
Quantity | 100 mg |
Content And Storage | -20°C |
Form | Lyophilized |
For Use With (Application) | Used as a stabilizer and/or blocker for EIA and RIA test systems |
Product Type | Purified Human gamma Globulin |
Product Content Correction
Product Title
MP Biomedicals™ γ-Globulin, Human Purified >