Learn More
MP Biomedicals™ Fat Diet, High, Modified, Paste
Fat Diet, High, Modified in paste form.
Brand:  MP Biomedicals™ 0296019210
Product Code. 11482931
  • 35750.00 NOK / 10kg
Explore more special offers
Description
Description
  • Not available in pellet form
  • Normal High Fat Diet (ICN96019210) modified to contain essential trace minerals, methionine and fiber
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Animal Diet
Paste
10 kg