Learn More
MP Biomedicals Choline Control Diet, Modified
Brand:  MP Biomedicals 0296041420
 View more versions of this product
Product Code. 11481482
  • 13410.00 NOK / 20kg
Explore more special offers
Specifications
Choline Control Diet
Pellet
Powder
15°C to 30°C
Vitamin Free Casein 10%, a-Protein 10%, Sucrose 51%, Alphacel, Non-Nutritive Bulk 5%, Lard 20%, Manganese Sulfates.H2O 150mg/kg diet, Zinc Chloride 20mg/kg diet, Chromium Potassium Sulfate.12H2O 55mg/kg diet, Sodium Selenite 1mg/kg diet, Salt Mixture Wesson 4%
To study liver injury in conjunction with MP's Methionine/Choline deficient diet
20,000 g