Learn More
MP Biomedicals™ Angiotensin-Converting Enzyme Substrate
Free acid
Brand:  MP Biomedicals™ 0215274790
Product Code. 11473901
  • 7885.00 NOK / 500mg
Explore more special offers
Description
Description
Hippuryl-His-Leu (Benzoyl-Gly-His-Leu)
  • A fluorogenic substrate for angiotensin-converting enzyme
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Off-White
-20°C
Angiotensin-converting Enzyme Substrate
Powder
Continuous spectrophortometric assays of Angiotensin Converting Enzyme
500 mg
399.4
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
MP Biomedicals™ Angiotensin-Converting Enzyme Substrate > 500mg