Learn More
MP Biomedicals AIN-76 Semipurified Diet
Rat or mouse
Brand:  MP Biomedicals 0290545220
 View more versions of this product
Product Code. 11492761
  • 9070.00 NOK / 20kg
Explore more special offers
Description
Description
American Institute of Nutrition
TRUSTED_SUSTAINABILITY
Specifications
Specifications
AIN-76 Semipurified Diet
Powder
Powder
Rat, Mouse
Casein Purified High Nitrogen 20%, DL-Methionine 0.3%, Sucrose 50%, Corn Starch 15%, Alphacel, Non-Nutritive Bulk 5%, Corn Oil 5%, Choline Bitartrate 0.2%, AIN Mineral Mix 3.5%, AIN Vitamin Mix 1%
In-vitro translation experiments and in biochemistry to isolate proteins
20,000 g
15°C to 30°C