Learn More
MP Biomedicals AIN-76 Semipurified Diet
Rat or mouse
Brand: MP Biomedicals 0290545250
This item is not returnable.
View return policy
Description
American Institute of Nutrition
Specifications
AIN-76 Semipurified Diet | |
Powder | |
Powder | |
Rat, Mouse |
Casein Purified High Nitrogen 20%, DL-Methionine 0.3%, Sucrose 50%, Corn Starch 15%, Alphacel, Non-Nutritive Bulk 5%, Corn Oil 5%, Choline Bitartrate 0.2%, AIN Mineral Mix 3.5%, AIN Vitamin Mix 1% | |
In-vitro translation experiments and in biochemistry to isolate proteins | |
50,000 g | |
15°C to 30°C |