missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse S100A8 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 17 publications
Brand: R&D Systems AF3059
This item is not returnable.
View return policy
Description
S100A8 Polyclonal specifically detects S100A8 in Mouse samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.
Specifications
S100A8 | |
Polyclonal | |
Unconjugated | |
P27005 | |
S100A8 | |
E. coli-derived recombinant mouse S100A8 Pro2-Glu89 Accession # P27005 | |
100 μg | |
Primary | |
Detects mouse S100A8 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant mouse (rm) S100A9, rmS100A10, and recombinant human S100B is observed. | |
Mouse | |
IgG |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 0.25 ug/mL, Simple Western 2.5 ug/mL, Immunocytochemistry 5-15 ug/mL | |
60B8AG, CAGACP-10, calgranulin A, Calgranulin-A, Calprotectin L1L subunit, CFAG, CFAGL1Ag, CGLA, Cystic fibrosis antigen, Leukocyte L1 complex light chain, MA387, MIF, Migration inhibitory factor-related protein 8, MRP8, MRP-8, MRP8S100 calcium binding protein A8 (calgranulin A), NIF, P8, protein S100-A8, S100 calcium binding protein A8, S100 calcium-binding protein A8, S100 calcium-binding protein A8 (calgranulin A), Urinary stone protein band A | |
Goat | |
Affinity Purified | |
RUO | |
6279 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Mouse S100A8 Antibody, R&D Systems™ > 100μg; Unlabeled
Spot an opportunity for improvement?Share a Content Correction