Learn More
R&D Systems™ Mouse Mesothelin (NP_061345.1) VersaClone cDNA
Brand:  R&D Systems™ RDC3306
Product Code. 18702177
  • 7505.00 NOK / 10µg
Explore more special offers
Specifications
Mesothelin
10232
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
10 μg
NP_061345.1
CAK1, CAK1 antigen, megakaryocyte potentiating factor, mesothelin, MPF, MPFSMRP, MSLN, Pre-pro-megakaryocyte-potentiating factor, SMR, soluble MPF mesothelin related protein
Mouse