missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse FOLR1 Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF6936
This item is not returnable.
View return policy
Description
FOLR1 Polyclonal specifically detects FOLR1 in Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry.
Specifications
FOLR1 | |
Polyclonal | |
Unconjugated | |
P35846 | |
FOLR1 | |
Chinese hamster ovary cell line CHO-derived recombinant mouse FOLR1 Arg23-Ser232 Accession # P35846 | |
100 μg | |
Primary | |
Detects mouse FOLR1 in direct ELISAs. In direct ELISAs, approximately 30% cross-reactivity with recombinant human FOLR1 is observed. | |
Mouse | |
IgG |
Immunocytochemistry | |
LYOPH | |
Immunocytochemistry 5-15 ug/mL | |
Adult folate-binding protein, FBP, folate binding protein, Folate receptor 1, folate receptor 1 (adult), folate receptor alpha, Folate receptor, adult, Folbp1, FOLR, FR-alpha, KB cells FBP, MOv18, Ovarian tumor-associated antigen MOv18 | |
Sheep | |
Affinity Purified | |
RUO | |
2348 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Mouse FOLR1 Antibody, R&D Systems™ > 100μg; Unlabeled
Spot an opportunity for improvement?Share a Content Correction