missing translation for 'onlineSavingsMsg'
Learn More

Mouse anti-Chicken Ig Mix Secondary Antibody, Invitrogen™

Mouse Monoclonal Secondary Antibody

4075.00 NOK

Specifications

Antigen Chicken Ig Mix
Clone CIG10-196.101
Applications ELISA
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11584990
View Documents
Invitrogen
SA110644
500 μg
4075.00 NOK
1mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SA1-10644 detects chicken Immunoglobulin. This antibody does not cross-react with immunoglobulins from bovine, canine, feline, ovine, porcine, equine, rabbit, human, donkey, goat, or guinea pig. SA1-10644 has been successfully used in ELISA applications. The SA1-10644 immunogen is purified chicken immunoglobulin.

Anti-Chicken secondary antibodies are affinity-purified antibodies with well-characterized specificity for the chicken immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications

Specifications

Chicken Ig Mix
ELISA
Unconjugated
Mouse
Chicken
Purified chicken immunoglobulin.
Secondary
-20° C, Avoid Freeze/Thaw Cycles
CIG10-196.101
Monoclonal
Liquid
RUO
PBS with no preservative; pH 7.4
IgG1
Protein A
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.