Learn More
R&D Systems™ Mouse Adenosine A2b R (NP_031439) VersaClone cDNA
R&D Systems™ Mouse Adenosine A2b R (NP_031439) VersaClone cDNA is an adenosine receptor that is a member of the G proteincoupled receptor superfamily.
Brand:  R&D Systems™ RDC2227
Product Code. 18715179
  • 3335.00 NOK / 10µg
Explore more special offers
Description
Description
The Mouse Adenosine A2b R (NP_031439) VersaClone cDNA clone contains the complete ORF for Adenosine A2b R, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Adenosine A2b R insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
mAdora2b
10 μg at 0.2 μg/μL
1012bp
136
Mouse
NP_031419
cDNA is provided in 10 mM Tris-Cl
8.5
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.