Learn More
R&D Systems™ Mouse Adenosine A2b R (NP_031439) VersaClone cDNA
R&D Systems™ Mouse Adenosine A2b R (NP_031439) VersaClone cDNA is an adenosine receptor that is a member of the G proteincoupled receptor superfamily.
Brand: R&D Systems™ RDC2227
This item is not returnable.
View return policy
Description
The Mouse Adenosine A2b R (NP_031439) VersaClone cDNA clone contains the complete ORF for Adenosine A2b R, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Adenosine A2b R insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Specifications
mAdora2b | |
10 μg at 0.2 μg/μL | |
1012bp | |
136 | |
Mouse |
NP_031419 | |
cDNA is provided in 10 mM Tris-Cl | |
8.5 | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
10 μg |