Learn More
Motic™ Moticam X5 Plus Camera
The Moticam X5 Plus is a high resolution streaming WiFi camera. By creating its own WiFi signal, simply connect with your Android or iOS device and view, capture, or edit live images from your microscope with our free MotiConnect app.
Brand: Motic™ 1100600101322
Description
- C-mount for lens
- MotiConnect software is available for free download (with free updates included)
- Live Display Modes: 1920 x 1080 px, 1280 x 720 px
Specifications
WiFi Camera | |
CMOS | |
30 fps |
CS Ring Adapter, Focusable Lens, 30 mm and 38 mm Eyepiece Adapters, USB 2.0 Cable, Motic™ 4-dot Calibration Slide, Macro Tube, Motic™ Images Plus 3.0 PC/OSX/Linux | |
1/3 in. |