missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MORF4L1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55257-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody has no cross-reaction to MORF4L2.
Cellular senescence is the terminal non-dividing state that normal cells enter following completion of their proliferative potential. Fusions of immortal human cell lines with each other have led to their assignment to one of four complementation groups. Mortality factor 4 (MORF4) was identified as the lead member of a family of transcription factor-like proteins that reverses this immortal phenotype. Like other members in this family, MORF4 is localized to the nucleus and possesses transcription factor-like motifs such as helix-loop-helix and a leucine zipper motif that might allow it to form transcriptionally active homo- or heterodimers. MORF4 has been shown to bind to the transcription corepressors mSin3A and TLE, suggesting that together, these complexes may play a role in transcriptional repression of genes that lead to cellular senescence.Specifications
MORF4L1 | |
Polyclonal | |
Unconjugated | |
MORF4L1 | |
Eaf3, FWP006, HSPC008, HSPC061, HsT17725, MORF related gene 15 protein, MORF4L1, MORFRG15, mortality factor 4 like 1, MRG15, PP368, Protein MSL3 1, S863 6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10933, 21761, 300891 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.55 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P60762, Q6AYU1, Q9UBU8 | |
Morf4l1 | |
Peptide | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction