missing translation for 'onlineSavingsMsg'
Learn More

Molybdic Acid Dihydrate Sodium Salt, (Crystalline),MP Biomedicals

Product Code. 11412452 Shop All MP Biomedicals Products
100g
Click to view available options
100g
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 10102-40-6
Physical Form Crystalline
Quantity 100 g
Molecular Formula MoNa2O4
MDL Number MFCD00149170
Synonym sodium molybdate dihydrate, sodium molybdate vi dihydrate, disodium molybdate dihydrate, unii-8f2sxi1704, na2moo4.2h2o, disodium dihydrate molybdate, molybdic acid sodium salt dihydrate, molybdic acid, disodium salt, dihydrate, sodium molybdate 2 hydrate, dipotassium dihydrate molybdate
InChI Key TVXXNOYZHKPKGW-UHFFFAOYSA-N
SMILES [Na+].[Na+].[O-][Mo]([O-])(=O)=O
IUPAC Name disodium dioxomolybdenumbis(olate)
Molecular Weight (g/mol) 205.93
PubChem CID 16211258
ChEBI CHEBI:75213
Formula Weight 241.9
Chemical Name or Material Molybdic Acid Dihydrate Sodium Salt
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Molybdic Acid Dihydrate Sodium Salt, (Crystalline),MP Biomedicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.