Learn More
Invitrogen™ Molecular Probes™ Press-to-Seal™ Silicone Isolator
For versatility and flexibility, isolators can be easily trimmed to custom fit enclosures
Brand: Invitrogen™ P18175
Additional Details : Weight : 0.00450kg
Description
- Available either precut to specific interior dimensions
- Isolators are autoclavable and noncytotoxic
- Can be used to isolate cells in culture dishes or to separate specimens on microscope slides
Cell Analysis, Cellular Imaging, IHC Sample Preparation, IHC Staining and Detection, Immunocytochemistry (ICC), Immunofluorescence (IF), Immunofluorescence Sample Preparation, Immunohistochemistry (IHC)
Specifications
Press-to-Seal™ Silicone Sheet | |
1 | |
1 well, 20mm dia., 1.0mm deep | |
20 mm |