Learn More
Invitrogen™ LIVE/DEAD™ Cell-Mediated Cytotoxicity Kit, for animal cells
Red and green stains used
Brand: Invitrogen™ L7010
2646.79 NOK valid until 2024-06-29
Use promo code "21620" to get your promotional price.
Additional Details : Weight : 0.01000kg
Description
The LIVE/DEAD Cell-Mediated Cytotoxicity Kit measures natural killer (NK) cell-mediated, lymphokine-activated killer (LAK) cell-mediated and T cell mediated cytotoxicity. Target cells are preincubated with the green-fluorescent membrane stain DiOC18 and then mixed with effector cells in the presence of the red-fluorescent, membrane-impermeant dye, propidium iodide. Live and dead target cells retain the green-fluorescent membrane stain; target and effector cells with compromised membranes exhibit red-fluorescent nucleic acid staining; live effector cells are nonfluorescent.
Cell Analysis, Cell Counting and Viability, Cell Counting, Viability, and Cryopreservation, Cell Culture, Cell Viability and Cytotoxicity, Cell Viability, Proliferation and Function, Flow Cytometry, Flow Cytometry Viability and Cytotoxicity Assays, Flow Cytometry of Cellular Processes
Order Info
Shipping Condition: Room temperature
Specifications
Mammalian Cells, Eukaryotic Cells | |
Store in refrigerator at 2°C to 8°C and protect from light. |
|
Fluorescence | |
Fluorescence Microscope, Flow Cytometer | |
Other Label(s) or Dye(s) | |
484, 536 nm | |
LIVE/DEAD™ |
Green, Red | |
LIVE/DEAD™ Cell-Mediated Cytotoxicity Kit, for animal cells | |
Viability Assay | |
Cell-Mediated Cytotoxicity Kit | |
501, 617 nm | |
Tube(s), Slide(s) | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.