missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP-2 Rabbit anti-Human, Mouse, Rat, Clone: 3Q7R10, Novus Biologicals™
Rabbit Monoclonal Antibody
1815.00 NOK - 4580.00 NOK
Specifications
Antigen | MMP-2 |
---|---|
Clone | 3Q7R10 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18304226
|
Bio-Techne
NBP3-15653-20UL |
20 μg |
1815.00 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18329485
|
Bio-Techne
NBP3-15653-100UL |
100 μg |
4580.00 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MMP-2 Monoclonal antibody specifically detects MMP-2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
MMP-2 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Angiogenesis, Apoptosis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, HIF Target Genes, Hypoxia, Metastasis, Tumor Suppressors | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
4313 | |
IgG | |
Affinity purified |
3Q7R10 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
72 kDa gelatinase, CLG4, CLG4A72 kDa type IV collagenase, collagenase type IV-A, EC 3.4.24, EC 3.4.24.24, Gelatinase A, matrix metallopeptidase 2 (gelatinase A, 72kDa gelatinase, 72kDa type IVcollagenase), matrix metalloproteinase 2 (gelatinase A, 72kD gelatinase, 72kD type IVcollagenase), Matrix metalloproteinase-2, matrix metalloproteinase-II, MMP-2, MMP-II, MONA, neutrophil gelatinase, TBE-1matrix metalloproteinase 2 (gelatinase A, 72kDa gelatinase, 72kDa type IVcollagenase) | |
A synthetic peptide corresponding to a sequence within amino acids 500-600 of human MMP-2 (P08253). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title