Learn More
MMAD-d8, MedChemExpress
MedChemExpress D8-MMAD is a deuterated form of MMAD, which is a microtubule disrupting agent.
Brand:  Medchem Express HY-15581S
Encompass_Preferred
 View more versions of this product
Product Code. 16401539
  • 17241.88 NOK / 1mg
Explore more special offers
Description
Description
D8-MMAD is a deuterated form of MMAD, which is a microtubule disrupting agent.
Specifications
Specifications
MMAD-d8
Off-White
C41H58D8N6O6S
DMSO : ≥ 100 mg/mL (128.35 mM)
779.11
99.12%
Cancer-programmed cell death
1 mg
Research
Demethyldolastatin 10-d8 Monomethylauristatin D-d8 Monomethyl Dolastatin 10-d8
O=C(N[C@H](C1=NC=CS1)CC2=CC=CC=C2)[C@H](C)[C@@H](OC)[C@H]3N(C(C[C@@H](OC)[C@H]([C@@H](C)CC)N(C([C@@](C([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])([2H])NC([C@H](C(C)C)NC)=O)=O)C)=O)CCC3
779.11
Research
Solid
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.