Learn More
Tocris Bioscience™ ML 297
Selective Kir3.1/3.2 (GIRK1/2) channel activator
Brand:  Tocris Bioscience™ 5380/10
1744.23 NOK valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18776638
  • 2205.00 NOK / 10mg
Explore more special offers
Description
Description
ML 297 is a selective Kir3.1/3.2 (GIRK1/2) channel activator (IC50 values are 160, 887 and 914 nM for GIRK1/2, GIRK1/4 and GIRK1/3 respectively). Exhibits no effect on GIRK2, GIRK2/3, Kir2.1 and Kv7.4 channels, and has minimal effect on a panel of other ion channels, receptors and transporters. Prevents PTZ -induced epileptic seizures in mice. Brain penetrant.
Specifications
Specifications
>99%
ML 297
ML 297
10 mg
328.32