Learn More
Tocris Bioscience™ (±)-ML 209
RORγt inverse agonist; suppresses Th17 cell differentiation
Brand: Tocris Bioscience™ 5987/50
6046.50 NOK valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
(±)-ML 209 is a RORγt inverse agonist (IC50 = 460 nM). Suppresses Th17 cell differentiation and IL17A expression in vitro. Does not suppress transcription of RORα. Minimal activity displayed on ERα, LXRα and thyroid hormone receptors (IC50 values >4.4 μM).Specifications
White | |
1334526-14-5 | |
441.52g/mol | |
Store at 4°C | |
50 mg |
Inhibitor | |
C25H31NO6 | |
Soluble to 100mM in DMSO | |
Agonist against RORγt |
For Laboratory Research Use Only