Learn More
ML-18, MedChemExpress
MedChemExpress ML-18 is a non-peptide bombesin receptor subtype-3 (BRS-3) antagonist with an IC50 of 4.8 μM.
Brand:  Medchem Express HY-101844
Encompass_Preferred
 View more versions of this product
Product Code. 16474696
  • 15835.35 NOK / 100mg
Explore more special offers
Description
Description
ML-18 is a non-peptide bombesin receptor subtype-3 (BRS-3) antagonist with an IC50 of 4.8 μM.
Specifications
Specifications
ML-18
Light Yellow
C32H35N5O5
DMSO : ≥ 100 mg/mL (175.55 mM)
569.65
98.44%
Solid
1422269-30-4
Research
100 mg
O=C([C@@H](NC(NC1=CC=C([N+]([O-])=O)C=C1)=O)CC2=CNC3=CC=CC=C23)NCC4(C5=CC=C(OC)C=C5)CCCCC4
569.65
Research
Cancer-Kinase/protease