Learn More
ML-18, MedChemExpress
MedChemExpress ML-18 is a non-peptide bombesin receptor subtype-3 (BRS-3) antagonist with an IC50 of 4.8 μM.
Brand:  Medchem Express HY-101844
Encompass_Preferred
 View more versions of this product
Product Code. 16426337
  • 1959.66 NOK / 5mg
Explore more special offers
Description
Description
ML-18 is a non-peptide bombesin receptor subtype-3 (BRS-3) antagonist with an IC50 of 4.8 μM.
Specifications
Specifications
ML-18
Light Yellow
C32H35N5O5
DMSO : ≥ 100 mg/mL (175.55 mM)
569.65
98.44%
Solid
1422269-30-4
Research
5 mg
O=C([C@@H](NC(NC1=CC=C([N+]([O-])=O)C=C1)=O)CC2=CNC3=CC=CC=C23)NCC4(C5=CC=C(OC)C=C5)CCCCC4
569.65
Research
Cancer-Kinase/protease
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.