Learn More
MK-8033, MedChemExpress
MedChemExpress MK-8033 is an orally active ATP competitive c-Met/Ron dual inhibitor (IC50s: 1 nM (c-Met),7 nM (Ron)), with preferential binding to the activated kinase conformation. MK-8033 can be used in the research of cancers, such as breast and bladder cancers, non-small cell lung cancers (NSCLCs).
Brand:  Medchem Express HY-13299
Encompass_Preferred
 View more versions of this product
Product Code. 16448327
  • 4899.16 NOK / 5mg
Explore more special offers
Description
Description
MK-8033 is an orally active ATP competitive c-Met/Ron dual inhibitor (IC50s: 1 nM (c-Met),7 nM (Ron)), with preferential binding to the activated kinase conformation. MK-8033 can be used in the research of cancers, such as breast and bladder cancers, non-small cell lung cancers (NSCLCs).
Specifications
Specifications
MK-8033
Light Yellow
C25H21N5O3S
DMSO : ≥ 46 mg/mL (97.55 mM)
471.53
Research
Cancer-Kinase/protease
1001917-37-8
Research
5 mg
O=S(NCC1=CC=CC=N1)(CC2=CC=C3C(C(C4=CC(C5=CN(N=C5)C)=CN=C4C=C3)=O)=C2)=O
471.53
Solid
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
MK-8033, MedChemExpress