Learn More
MK-8033, MedChemExpress
MedChemExpress MK-8033 is an orally active ATP competitive c-Met/Ron dual inhibitor (IC50s: 1 nM (c-Met),7 nM (Ron)), with preferential binding to the activated kinase conformation. MK-8033 can be used in the research of cancers, such as breast and bladder cancers, non-small cell lung cancers (NSCLCs).
Brand: Medchem Express HY-13299
Description
MK-8033 is an orally active ATP competitive c-Met/Ron dual inhibitor (IC50s: 1 nM (c-Met),7 nM (Ron)), with preferential binding to the activated kinase conformation. MK-8033 can be used in the research of cancers, such as breast and bladder cancers, non-small cell lung cancers (NSCLCs).Specifications
MK-8033 | |
Light Yellow | |
C25H21N5O3S | |
DMSO : ≥ 46 mg/mL (97.55 mM) | |
471.53 | |
Research | |
Cancer-Kinase/protease |
1001917-37-8 | |
Research | |
5 mg | |
O=S(NCC1=CC=CC=N1)(CC2=CC=C3C(C(C4=CC(C5=CN(N=C5)C)=CN=C4C=C3)=O)=C2)=O | |
471.53 | |
Solid |