Learn More
Mixed Standard, 10ppm, Reagecon™
Brand: Reagecon TOIC10
Quantity | 500 mL |
---|
This item is not returnable.
View return policy
Specifications
10 ppm | |
500 mL |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
TOIC1025D1 | Certificate of Analysis | 22/05/2025 | TOIC10 |
Product Content Correction
Product Title
Mixed Standard, 10ppm, Reagecon™ > Quantity: 500mL