missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kartell™ MiniFix Micropipettor
An innovative product line of fixed-volume air dispacement mini-micropipettes. Kartell™ MiniFix Micropipettor is designed for fast, repetitive, and precise operation and are competitively priced.
Brand: Kartell™ 0128600
237.96 NOK valid until 2025-04-13
Use promo code "24122" to get your promotional price.
This item is not returnable.
View return policy
Description
- Fully autoclavable
- Designed for fast, repetitive and precise operation
- Very compact in terms of size and weight
- Designed for routine or biological hazard work
- Kartell™ tips fit on the conical shaft as well as most other commercially available tips
- The plunger button has a different color based on dosing capacity
- Individually calibrated in compliance with ISO8655, the new international standard for piston-operated volumetric apparatus
- Carries the CE conformity mark
Specifications
±1.0% | |
Kartell™ 960/958 | |
Red | |
1 | |
10 μL | |
1/Pk. | |
10 g |
CE/IVD | |
Autoclavable | |
in Situ Analyses and Sampling, Routine or Biological Hazard Work | |
Micropipette | |
Routine or biological hazard work, in situ analyses and sampling | |
13 cm |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 12
Special Offers
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
Days
5Hours
15Minutes
22Seconds
32
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Kartell™ MiniFix Micropipettor > Volume Metric Range: 10μL
Spot an opportunity for improvement?Share a Content Correction