Learn More
Cole-Parmer Miniature Check Valves, Hose Barb, Polypropylene and Viton™
Versatile check valves can be installed in any position
42.45 NOK - 56.45 NOK
Product Code | Brand | Inlet | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Inlet | Price | Quantity & Availability | ||||||
30054373
|
Cole-Parmer
50115-48 |
1/4 in. hose barb |
56.45 NOK
Each |
Estimated Shipment: 16-01-2026
Log in to see stock. |
||||||
30054130
|
Cole-Parmer
50115-20 |
1/8 in. hose barb |
42.45 NOK
Each |
Estimated Shipment: 28-01-2026
Log in to see stock. |
||||||
30053078
|
Cole-Parmer
50115-21 |
3/16 in. hose barb |
56.45 NOK
Each |
Estimated Shipment: 16-01-2026
Log in to see stock. |
||||||
30054160
|
Cole-Parmer
50115-22 |
5/16 in. hose barb |
56.45 NOK
Each |
Estimated Shipment: 16-01-2026
Log in to see stock. |
||||||
Description
Prevent backflow to protect your equipment. Work with both liquids and gases. Use for pressure or vacuum applications. 0.18 psi breaking pressureSpecifications
Barbed | |
Viton |
Product Title
Cole-Parmer Miniature Check Valves, Hose Barb, Polypropylene and Viton™