Learn More
Axygen™ Mini Tube System
Polypropylene tube system for compound library storage
Brand:  Axygen™ MTS-12CP-C
 View more versions of this product
Product Code. 11346324
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Axygen™ Mini Tube Systems are available in 0.65 mL single mini tube or 1.1 mL single mini tube. They are also available in 8-cap or 12-cap strip format. Mini Tube System racks are compatible with Beckman Coulter, Tecan, and other automation workstations.
  • Rack features a one-way lid design and alpha-numeric grid
  • Molded-in condensation rings on the lid minimize cross contamination
TRUSTED_SUSTAINABILITY
Specifications
Specifications
12 Cap Strip
Polyethylene
Ensures positive sample identification
80 Strips/PK., 5 Packs/Cs.
Strip
Clear
Strip
80/Pk.
Non-sterile
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Axygen™ Mini Tube System > Non-sterile; Bulk, 12 cap strip