Learn More
Ohaus™ Mini Digital Vortex Mixer VXMNDG
OHAUS™ Mini Digital Vortex Mixer VXMNDG is built sturdy to provide a stable and reliable vortexing action.
Brand: Ohaus™ 30392124
This item is not returnable.
View return policy
Description
- Sturdy construction for stable and reliable vortexing action
- Ideal for applications that demand repeatable results
- Microprocessor control maintains set speed for strong, consistent mixing action
- Timer will display elapsed time or, when programmed to user defined time limit, the unit will shut off when time reaches zero
- Two modes of operation; continuous mode when using accessory attachments or touch mode which activates mixing when depressing the cup head
- LED displays for speed and time
Specifications
4.9 mm | |
320 V, 0.6 A, 50/60 Hz | |
21.00 x 12.30 cm | |
16.5 cm | |
21.00 cm | |
12.30 cm |
500 to 2500 rpm | |
0.6 A | |
6.50 in. | |
8.30 in. | |
4.80 in. | |
VXMNDG |
Promotions
-
Multiple Manufacturers
The Ultimate Lab Appliances Promotion
Enjoy discounts up to 78% on top-quality lab equipment and upgrade your laboratory with the best appliances at the lowest prices.Promo Code: 24075Expires: 2025-06-30
New Offer Save Up to 78%
Product Content Correction
Product Title
Ohaus™ Mini Digital Vortex Mixer VXMNDG > Dimensions (L x W x H): 21 x 12.3 x 16.5 cm