Learn More
Thermo Scientific™ Mini Capillary Unions
Thermo Scientific™ Mini Capillary Unions are for use with capillary GC columns.
Brand:  Thermo Scientific™ 290GU499
795.87 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10289074
  • 838.00 NOK / Each
Only null left
Estimated Shipment: 20-06-2025
Description
Thermo Scientific™ Mini Capillary Unions are for use with capillary GC columns.
Kit contains one union and five ferrules
Includes:
Union, 1; Ferrules, 5
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Capillary Union
For 0.53 mm I.D. Capillary
0.53 mm ID Capillary GC Column
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ Mini Capillary Unions > For 0.53mm I.D. Capillary