Learn More
Brady™ Mild Sheet Steel Portable Metal Group Lock Box
Simplifies lockout of large equipment
Brand: Brady™ 065699
954.75 NOK valid until 2025-08-24
Use promo code "24132" to get your promotional price.
This item is not returnable.
View return policy
Description
- Heavy steel lock box stores keys after machine or process is locked out
- Each authorized work team member adds a personal lock or tagout device on the group lock box
- Ensures that no one employee has access unless all employees have removed their locks or tags
- Accepts up to 13 individual locks
Specifications
Red | |
1 Ea. |
Promotions
-
Multiple Manufacturers
Everyday Lab Essentials
From wipes to waste bins, labels to liners – save on them all.Promo Code: 24132Expires: 2025-08-24
New Offer Shop Now
Product Content Correction
Product Title
Brady™ Mild Sheet Steel Portable Metal Group Lock Box > Portable metal lock box; Red