Learn More
Brady™ Mild Sheet Steel Lock Box
For group lockout situations involving a large number of workers and equipment, this Mild Sheet Steel Lock Box can offer a number of advantages over hasps.
Brand: Brady™ 045190
930.21 NOK valid until 2025-08-24
Use promo code "24132" to get your promotional price.
Description
- 13 lock holes on lid and can also be used as a lock storage box holding up to 40 padlocks.
- Slot in front allows keys to be inserted when box is locked.
- Dimensions: 88.9 x 226.06 x 152.4 mm (D x W x H)
Specifications
Lock Box | |
Blue | |
Portable Metal Group Lock Box | |
1 Ea. |
Promotions
-
Multiple Manufacturers
Everyday Lab Essentials
From wipes to waste bins, labels to liners – save on them all.Promo Code: 24132Expires: 2025-08-24
New Offer Shop Now