Learn More
Kinesis™ Microwave Vials
Brand:  Kinesis™ VMW02-C-100-SB
9258.38 NOK valid until 2025-06-13
Use promo code "24123" to get your promotional price.
Product Code. 15178102
  • 9820.00 NOK / Pack of 100
Price-Shattering Deals on Lab Glassware
Use promo code 24123 at checkout
View Available Promotions
Only null left
Estimated Shipment: 07-10-2025
Description
KX, for use with Initiator Systems (EXP only) from Biotage, contamination free 1st class hydrolytic borosilicate glass, resealable septa, 40°C to 300°C, 0.5mL to 2.0mL
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Kinesis™ Microwave Vials >