Learn More
Thermo Scientific™ Microtiter Tube Plug Strip
Seal 1.2mL microtiter tubes tightly for storage using these polyethylene plug strips.
Brand: Thermo Scientific™ 3425-11
248.02 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Seal 1.2mL microtiter tubes tightly for storage by using Thermo Scientific™ Microtiter Tube Plug Strips.
- LDPE non-autoclavable
- Eight-plug strips can be easily cut apart for individual samples
- Available sterile and nonsterile
Specifications
8-cap Strip | |
1.2mL Microtiter Tube 8-cap plug strip | |
Natural | |
120/Pk., 5/Cs. | |
Plug Strips |
Not Autoclavable | |
LDPE | |
10 sealed bags of twelve 8 cap strips/unit, 5 units/case | |
Sterile |