Learn More
Sampling Systems™ MicroThief™ Low Volume Powder Thief
Easily adjusted so that the operator can vary the size of the sample being taken. Sampling Systems™ MicroThief™ Low Volume Powder Thief is suitable for taking very small volume samples of free flowing powders and granules.
Brand:  Sampling Systems™ 1074A-500
 View more versions of this product
Product Code. 15859211
  • 13130.00 NOK / Each
Only null left
Estimated Shipment: 07-08-2025
Description
  • Made from high quality 316L stainless steel
  • Adjustable, takes a range of sample sizes
Specifications
Specifications
Stainless Steel (316L)
0.01 to 0.1 mL
5 mm
500 mm
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Sampling Systems™ MicroThief™ Low Volume Powder Thief > DepthMetric: 500mm