Learn More
Fisherbrand™ Microscope Adapter
Use this Microscope adapter with compound biological microscopes. Fisherbrand™ Microscope Adapters are ergonomically designed for everyday use.
3310.00 NOK
Specifications
Type | Microscope Adapter |
---|---|
Includes | Projection lens accessory |
Quantity | 1 Ea. |
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
11562653
|
Fisherbrand™
2757.2126 |
1/2 CCD Camera |
3310.00 NOK
Each |
Estimated Shipment: 16-04-2025
Log in to see stock. |
|||||
11552653
|
Fisherbrand™
2757.6132 |
1/3 CCD Camera |
3310.00 NOK
Each |
Estimated Shipment: 16-04-2025
Log in to see stock. |
|||||
Description
- Microscope Adapter
- Accessory for microscope models: FB69075 and FB69088
Specifications
Microscope Adapter | |
Projection lens accessory | |
1 Ea. |