Learn More
Simport™ Scientific Micromesh™ Biopsy Processing/Embedding Cassette in Quickload™ stacks
Secure specimens during processing with Simport Scientific™ Micromesh™ Biopsy Processing/Embedding Cassette. No need for biopsy pads thanks to the 1676 square 0.36mm openings of the mesh.
Description
In Quickload™ stacks
- 45° Anterior writing area
- 1676 square opening (0.36 mm)
- No biopsy pads needed
- 1 large 27 mm compartment
- Recessed cover
- Suitable for automated printers
- Made of acetal
- Resistant to the chemical action of most histological solvents
Order Info
Each case contains 50 Quickload™ stacks of 40 cassettes.
Certifications
CE-IVD
Specifications
Acetal | |
2000 |
Product Title
Simport™ Scientific Micromesh™ Biopsy Processing/Embedding Cassette in Quickload™ stacks