Learn More
Ansell Microgard™ 2000 Standard White Coveralls with Collar Product Code.: 15576504
Add to basket
Ansell Microgard™ 2000 Standard White Coveralls with Collar
Product Code. 15576504 Shop All Ansell Products
Small
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Product Code. 15576504
Brand: Ansell WH20B0010302
Add to basket
Add to basket
Allows perspiration to escape from the suit yet withstands saturation of liquid chemicals. Ansell Microgard™ 2000 Standard White Coveralls with Collar filter 100% of particulates down to 0.01 microns in size.
  • Coverall with collar
  • White bound seams
  • Elasticated hood, waist, wrists and ankles
  • Includes fingerloops
  • 2-way front zipper with re-sealable storm flap.
  • Excellent liquid penetration resistance and barrier to fine particulates (>0.01 microns)
  • High quality two-way stretch microporous film provides an effective liquid and particle barrier
  • Moisture vapor permeable to help reduce the risk of heat stress
  • Silicone free - Critical in paint spraying applications
  • Low linting - Reduces the risk of fibre contamination in some critical areas
  • Optimised body fit provides wearer comfort and safety
  • Anti-static treatment
TRUSTED_SUSTAINABILITY
Specifications
Product Type General Purpose Coveralls
Size Small
Gender Unisex
Material Microporous Polyethylene Film
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Closure Type Front Zipper with Storm Flap
Seam Type Bound
Sterility Non-sterile
Certifications/Compliance (Europe) Cat III Type 5-B, 6-B, EN1073-2, EN14126, EN1149-1, DIN 32781
Model WH20-B-00-103
No. of Pockets None
Show More Show Less
Product Content Correction
Product Title
Ansell Microgard™ 2000 Standard White Coveralls with Collar >